Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPTLC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521141
Description
A suggested positive control is human lung tissue lysate. PA5-21141 can be used with blocking peptide PEP-1255.
Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.
Specifications
SPTLC1 | |
Polyclonal | |
Unconjugated | |
Sptlc1 | |
AW552086; C77762; E030036H05; fc75h04; fp41c08; hLCB1; HSAN; HSAN1; HSN1; hypothetical protein LOC553981; hypothetical protein LOC614165; LBC1; LCB 1; LCB1; long chain base biosynthesis protein 1; MGC14645; rCG_44191; RGD1306617; RP11-118F2.1; serine C-palmitoyltransferase; serine palmitoyltransferase 1; serine palmitoyltransferase long chain base subunit 1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; serine-palmitoyl-CoA transferase 1; SPT 1; Spt1; SPTI; sptlc1; wu:fc75h04; wu:fp41c08; zgc:112247 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10558 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O15269 | |
Sptlc1 | |
A 17 amino acid peptide near the carboxy terminus of human SPT1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction