Learn More
Cayman ChemicalSupplier Diversity Partner SQ 29,548, 98672-91-4, 1 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 190251MG
Selective TXA2 receptor antagonist (Ki = 4.1 nM); inhibits U-46619-induced platelet aggregation (IC50 = 0.06 µM)
Catalog No. NC0162195