Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SQSTM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA595255
Description
Synthetic peptide sequence: 69-96aa, DEDGDLVAFSSDEELTMAMSYVKDDIFR. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.Specifications
SQSTM1 | |
Polyclonal | |
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl, 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide | |
O08623, Q13501, Q64337 | |
SQSTM1 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human SQSTM1/p62, identical to the related mouse and rat sequences. | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Unconjugated | |
SQSTM1 | |
Sequestosome-1; EBI3-associated protein of 60 kDa; EBIAP; p60; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; Ubiquitin-binding protein p62; SQSTM1; ORCA, OSIL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
113894, 18412, 8878 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title