Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalane, 99%, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass bottle
111-01-3
C30H62
422.826
PRAKJMSDJKAYCZ-UHFFFAOYSA-N
squalane,perhydrosqualene,dodecahydrosqualene,cosbiol,vitabiosol,spinacane,robane,hexamethyltetracosane,tetracosane, 2,6,10,15,19,23-hexamethyl,hexamethyl tetracosane
8089
2,6,10,15,19,23-hexamethyltetracosane
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
111-01-3
C30H62
422.826
PRAKJMSDJKAYCZ-UHFFFAOYSA-N
squalane,perhydrosqualene,dodecahydrosqualene,cosbiol,vitabiosol,spinacane,robane,hexamethyltetracosane,tetracosane, 2,6,10,15,19,23-hexamethyl,hexamethyl tetracosane
8089
2,6,10,15,19,23-hexamethyltetracosane
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Acidity 0.2mg KOH/g max.
Saponification Value 1mg KOH/g max.
Density 0.8085g/mL
Quantity 2.5L
Packaging Glass bottle
Refractive Index 1.4510 to 1.4525 (20°C, 589nm)
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Formula Weight 422.82
Physical Form Liquid
Percent Purity ≥98.5% (GC)
Chemical Name or Material Squalane
Show More Show Less