Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalene, 99+%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
111-02-4
C30H50
410.73
MFCD00008912
YYGNTYWPHWGJRM-AAJYLUCBSA-N
squalene,trans-squalene,spinacene,supraene,all-trans-squalene,2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene,e,e,e,e-squalene,unii-7qwm220fjh,7qwm220fjh,spinacen
638072
CHEBI:15440
(6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
CC(=CCCC(=CCCC(=CCCC=C(C)CCC=C(C)CCC=C(C)C)C)C)C
111-02-4
C30H50
410.73
MFCD00008912
YYGNTYWPHWGJRM-AAJYLUCBSA-N
squalene,trans-squalene,spinacene,supraene,all-trans-squalene,2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene,e,e,e,e-squalene,unii-7qwm220fjh,7qwm220fjh,spinacen
638072
CHEBI:15440
(6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
CC(=CCCC(=CCCC(=CCCC=C(C)CCC=C(C)CCC=C(C)C)C)C)C
Quantity 500mL
Infrared Spectrum Authentic
Saponification Value 1mg KOH/g max.
Color Colorless to Yellow
Acidity 1mg KOH/g max.
Packaging Glass bottle
Density 0.855g/mL
Refractive Index 1.4945 to 1.4965 (20°C, 589nm)
Assay Percent Range 99+%
Linear Formula {(CH3)2C{=CHCH2CH2C(CH3)}2=CHCH2-}2
Formula Weight 410.71
Physical Form Oil
Percent Purity ≥99.0%
Chemical Name or Material Squalene
Show More Show Less