Learn More
Eppendorf™ Square Buckets for Eppendorf™ Four-Place Rotor
For use with 05-400-328 to accept round-bottom tubes from 5 to 25mL with appropriate adapters
Manufacturer:  Eppendorf™ 022639307
Catalog No. 05-400-309
Add to cart
Specifications
Specifications
A-4-38 swing bucket rotor for 5702/5702R/5702RH centrifuge