Learn More
Eisco™ Squibb Separatory Funnels with PTFE Key Stopcock
SCI_ED
High quality borosilicate 3.3 glass funnel with PTFE key stopcocks and interchangeable polypropylene stoppers
$32.60 - $65.80
Specifications
Material Borosilicate Glass
Description Ungraduated
Graduated No
Quantity 1
Product Type Squibb Separatory Funnel
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Stopper Number Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Stopper Number Price Quantity & Availability  
S89286
SCI_ED
View Documents
Eisco™
CH0479A
50 mL 14/23
Each for $35.80
Only null left
Add to Cart
S89287
SCI_ED
View Documents
Eisco™
CH0479B
100 mL 19/26
Each for $32.60
Only null left
Add to Cart
S89288
SCI_ED
View Documents
Eisco™
CH0479C
250 mL 19/26
Each for $39.25
Only null left
Edge
Add to Cart
S24520
SCI_ED
View Documents
Eisco™
CH0479D
500 mL 24/29
Each for $40.55
Only null left
Edge
Add to Cart
S89289
SCI_ED
View Documents
Eisco™
CH0479E
1000 mL 29/32
Each for $50.90
Only null left
Add to Cart
S89291
SCI_ED
View Documents
Eisco™
CH0479F
2000 mL 29/32
Each for $65.80
Only null left
Add to Cart
Description
Description
  • Pear shaped separatory funnel
  • Made from high quality borosilicate 3.3 glass
  • Ungraduated
  • PTFE key stopcock
  • Includes interchangeable, chemical resistant polypropylene stopper
  • Used to separate different liquids
  • Funnel and stopper both autoclavable
  • Warranty and Services
    1 year limited manufacturer warranty
    Specifications
    Specifications
    Borosilicate Glass
    No
    Squibb Separatory Funnel
    Ungraduated
    1
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.