Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3110 Dual Carabiner

Squids™ 3110 Dual Carabiner tethers tools to help worker prevent injury, damage, and lost productivity from dropped tools.

Specifications

Load Bearing Capacity (English) 10 lb.
Load Bearing Capacity (Metric) 4.5 kg
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-820-298
View Documents
Ergodyne™
19023
Standard N/A N/A N/A
19-820-300
View Documents
Ergodyne™
19033
Extended N/A N/A N/A
Description

Description

  • Energy absorbing cord with tubular nylon webbing
  • Single action aluminum carabiner
  • Captive eye carabiner
  • Reflective accent for higher visibility
  • Lock stitching at both ends for extra security
  • Certified to a 2:1 safety factor
Specifications

Specifications

10 lb.
4.5 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.