Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3740 Hand Tool Attachment Trap Slips™

Squids™ Slips™ are retrofit tool attachments for small hand tools, which help reduce the risk of dropped objects and increase workplace safety.

$18.87 - $22.45

Specifications

Certifications/Compliance ANSI/ISEA 121-2018 standard
Color Orange
Load Bearing Capacity (English) 1 lb.
Load Bearing Capacity (Metric) 0.45 kg
View More Specs

Products 9
Catalog Number Mfr. No. Includes Size Price Quantity & Availability  
Catalog Number Mfr. No. Includes Size Price Quantity & Availability  
19-820-339
View Documents
Ergodyne™
19740
4/Pk. Small
Each for $18.87
Add to Cart
 
19-820-340
View Documents
Ergodyne™
19741
4/Pk. Medium
Each for $21.20
Add to Cart
 
19-820-341
View Documents
Ergodyne™
19742
4/Pk. Large
Each for $21.20
Add to Cart
 
19-820-342
View Documents
Ergodyne™
19743
4/Pk. X-Large
Each for $22.45
Add to Cart
 
19-820-343
View Documents
Ergodyne™
19744
4/Pk. Variety
Each for $21.95
Add to Cart
 
19-820-378
View Documents
Ergodyne™
19940BULK
60/Pk. Small Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-820-379
View Documents
Ergodyne™
19941BULK
60/Pk. Medium Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-820-380
View Documents
Ergodyne™
19942BULK
60/Pk. Large Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-820-381
View Documents
Ergodyne™
19943BULK
60/Pk. X-Large Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Approved to the ANSI/ISEA 121-2018 standard
  • One-piece design easily slides onto small hand tools without requiring additional tools to install
  • Swiveling exterior ring spins 360 degrees, allowing screwdrivers and other rotating tools to be used without issue
  • ABS outer ring provides strength and durability in all environments
  • Flexible inner core grips tool after application
  • Intended for singled use
Specifications

Specifications

ANSI/ISEA 121-2018 standard
Orange
1 lb.
0.45 kg
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit