Learn More
Sqwincher™ Activity Drinks, Powder Concentrate
Thirst-quenching drink that's low in sodium and high in potassium
Manufacturer:  Sqwincher CorporationSupplier Diversity Partner 016004OR
 View more versions of this product
Catalog No. 19-819-541
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Orange
80 Pk./Cs.
9.53 oz.
Provides supplemental source of energy, prevents or reduces severity of heat-stress disorders
Powder Pack
0.28L