Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR 1001, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1335106-03-0
C15H13F6N3O4S2
477.396
OZBSSKGBKHOLGA-UHFFFAOYSA-N
n-4-methyl-5-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl amino sulfonyl-2-thiazolylacetamide,sr hplc,n-5-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl sulfamoyl-4-methyl-1,3-thiazol-2-yl acetamide
44241473
N-[5-[[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]sulfamoyl]-4-methyl-1,3-thiazol-2-yl]acetamide
CC1=C(SC(=N1)NC(=O)C)S(=O)(=O)NC2=CC=C(C=C2)C(C(F)(F)F)(C(F)(F)F)O
This item is not returnable. View return policy
1335106-03-0
C15H13F6N3O4S2
477.396
OZBSSKGBKHOLGA-UHFFFAOYSA-N
n-4-methyl-5-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl amino sulfonyl-2-thiazolylacetamide,sr hplc,n-5-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl sulfamoyl-4-methyl-1,3-thiazol-2-yl acetamide
44241473
N-[5-[[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]sulfamoyl]-4-methyl-1,3-thiazol-2-yl]acetamide
CC1=C(SC(=N1)NC(=O)C)S(=O)(=O)NC2=CC=C(C=C2)C(C(F)(F)F)(C(F)(F)F)O

RORα/γ inverse agonist

Retinoic acid receptor-related orphan receptor (ROR) α and γ inverse agonist (Ki values are 172 and 111 nM respectively). Demonstrates no activity at RORβ or LXR. Suppresses Th17 cell differentiation and cytokine expression and reduces the severity of disease in an animal model of mulitple sclerosis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 477.4
Percent Purity >98%
Chemical Name or Material SR 1001

Recommended Storage : Store at +4°C