Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 13800 Available on GSA/VA Contract for Federal Government customers only.

Potent MCT1 inhibitor

$209.50 - $859.00

Specifications

Chemical Name or Material 2,6-Dihydro-7-[(3-hydroxypropyl)thio]-2-methyl-4-(2-methylpropyl)-6-(1-naphthalenylmethyl)-1H-pyrrolo[3,4-d]pyridazin-1one
Molecular Formula C25H29N3O2S
CAS 227321-12-2
Product Type Target: Monocarboxylate Transporter Inhibitors
Formula Weight 435.58
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
543110 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5431/10
10mg Each for $209.50
Add to cart
 
543150 View Documents Tocris Bioscience™
5431/50
50mg Each for $859.00
Add to cart
 
Description

Description

Potent MCT1 inhibitor (IC50 = 0.5 nM); blocks proliferation of Raji lymphoma cells in vitro. Also inhibits lactate uptake in breast cancer cells in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

2,6-Dihydro-7-[(3-hydroxypropyl)thio]-2-methyl-4-(2-methylpropyl)-6-(1-naphthalenylmethyl)-1H-pyrrolo[3,4-d]pyridazin-1one
227321-12-2
435.58
C25H29N3O2S
Target: Monocarboxylate Transporter Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit