Learn More
Tocris Bioscience™ SR 141716A
Selective CB1 inverse agonist
$108.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09-231-0R View Documents Tocris Bioscience™
0923/10
10mg Each for $108.00
Add to cart
09-235-0 View Documents Tocris Bioscience™
0923/50
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Potent and selective cannabinoid CB1 receptor antagonist (Ki = 1.98 nM). Also acts as an inverse agonist reversing adenylyl cyclase inhibition by WIN 55,212-2 (Cat.No. 1038) (IC50 = 48 nM). Displays no activity at CB2 receptors. Reduces food intake and body weight in orally dosed non-obese Wistar rats. Also μ-opioid receptor antagonist (Ki = 652 nm)
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at RT