Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 141716A

Selective CB1 receptor antagonist

Manufacturer:  Tocris Bioscience™ 0923/50

 View more versions of this product

Catalog No. 09-235-0


Add to cart

Description

Description

Potent and selective cannabinoid CB1 receptor antagonist (Ki = 1.98nM). Also acts as an inverse agonist reversing adenylyl cyclase inhibition by WIN 55,212-2 (Cat.No. 1038) (IC50 = 48nM). Displays no activity at CB2 receptors. Reduces food intake and body weight in orally dosed non-obese Wistar rats.
Specifications

Specifications

N-(Piperidin-1-yl)-5-(4-chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide hydrochloride
>99%
Observed MW: 500.25
Soluble to 100mM in DMSO
158681-13-1
C22H21Cl3N4O.HCl
50mg
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at RT

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit