Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR 2640 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
146662-42-2
C23H19ClN2O3
406.866
KDSYWFCTUKABKE-UHFFFAOYSA-N
sr 2640 hydrochloride,2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride,2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride,2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride,benzoic acid,2-3-2-quinolinylmethoxy phenyl amino,2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
45073453
2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl
This item is not returnable. View return policy
146662-42-2
C23H19ClN2O3
406.866
KDSYWFCTUKABKE-UHFFFAOYSA-N
sr 2640 hydrochloride,2-3-2-quinolinylmethoxy phenyl amino benzoic acid hydrochloride,2-3-quinolin-2-ylmethoxy phenyl amino benzoic acid hydrochloride,2-3-2-quinolinylmethoxy phenyl amino benzoic acidhydrochloride,benzoic acid,2-3-2-quinolinylmethoxy phenyl amino,2-3-2-quinolinylmethoxy phenyl a??mino benzoic acid hydrochloride
45073453
2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
C1=CC=C2C(=C1)C=CC(=N2)COC3=CC=CC(=C3)NC4=CC=CC=C4C(=O)O.Cl

Potent, selective LTD4/LTE4 receptor antagonist

Potent and selective competitive leukotriene D4 and E4 receptor antagonist. Inhibits LTD4- but not histamine-induced guinea pig ileum and trachea contraction (pA2 = 8.7). Also inhibits LTD4-attenuation of human PMN chemotaxis. Orally active in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 406.87
Percent Purity >99%
Chemical Name or Material SR 2640 hydrochloride

Recommended Storage : Desiccate at Room Temperature