Tocris™ SR 48692

Selective non-peptide NTS1 antagonist

Manufacturer: Tocris™ 3721/10

 View more versions of this product

Catalog No. 37-211-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors SR 48692
    Purity >98
    Molecular Weight 587.07
    Quantity 10mg
    Product Type SR 48692

    Neurotensin antagonist; selective for NTS1 over NTS2 (apparent affinity, Ke, is 36nM for NTS1). Competitively inhibits binding of [125I]-neurotensin to HT29 and N1E115 cell membranes (IC50 values are 15.3 and 20.4nM respectively). Orally bioavailable.

    • Chemical Name: 2-[[[1-(7-Chloro-4-quinolinyl)-5-(2,6-dimethoxyphenyl)-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13,7]decane-2-carboxylic acid
    • Formula: C32H31ClN4O5
    • CAS Number: 146362-70-1
    • Solubility: Soluble to 20mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit