Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 9238

Potent and selective LXR inverse agonist

$164.90 - $678.50

Specifications

Chemical Name or Material Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate
Molecular Formula C31H33NO7S2
CAS 1416153-62-2
Product Type Target: LXR-like Receptor Antagonists
Formula Weight 595.73
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
585410 View Documents Tocris Bioscience™
5854/10
10mg Each for $164.90
Add to cart
 
585450 View Documents Tocris Bioscience™
5854/50
50mg Each for $678.50
Add to cart
 
Description

Description

Potent and selective LXR inverse agonist (IC50 values are 43 and 214 nM for LXRβ and LXRα, respectively). Selectively inhibits liver LXR over peripheral LXR. Exhibits selectivity for LXR over a panel of 20 other nuclear receptors, including FXR. Reduces hepatic steatosis in obese mice.
Specifications

Specifications

Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate
1416153-62-2
595.73
C31H33NO7S2
Target: LXR-like Receptor Antagonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit