Learn More
Tocris Bioscience™ SR 9243
Selective LXR inverse agonist
$159.90 - $668.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
61-381-0 View Documents Tocris Bioscience™
6138/10
Each for $159.90
Add to cart
61-385-0 View Documents Tocris Bioscience™
6138/50
Each for $668.50
Add to cart
Description
Description
Selective LXR inverse agonist. Exhibits no significant activity at other nuclear receptors at maximally effective concentration. Inhibits the Warburg effect and lipogenesis by down-regulation of LXR-mediated gene expression. Selectively reduces cell viability and induces apoptosis in cancer cell lines in vitro. Suppresses hepatic steatosis and inhibits growth of tumor xenografts in mice.