Learn More
SRBC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302419AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein kinase C delta (PKC-d) binding protein (PRKCDBP), also known as cavin-3, is a member of the cavin family of proteins that are involved in caveolin formation and regulation. PRKCDBP was initially identified in a screen of cultured cell lines for proteins that were strongly induced by serum starvation. Studies indicate that PRKCDBP binds not only to PKC-delta but also to caveolin-1 and helps regulate caveolin traffic and function. Similar to other members of the cavin family, the expression of PRKCDBP was found to be down-regulated in various cancer cell lines, suggesting a possible tumor suppressor function of PRKCDBP.Specifications
SRBC | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q969G5 | |
CAVIN3 | |
Between 250 and 261 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CAVIN3 | |
PRKCDBP; protein kinase C, delta binding protein; HSRBC; MGC20400; SRBC; sdr-related gene product that binds to c-kinase; cavin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112464 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.