Learn More
SRBC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | SRBC |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2536
![]() |
Bethyl Laboratories, Inc
A302418AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
The protein kinase C delta (PKC-d) binding protein (PRKCDBP), also known as cavin-3, is a member of the cavin family of proteins that are involved in caveolin formation and regulation. PRKCDBP was initially identified in a screen of cultured cell lines for proteins that were strongly induced by serum starvation. Studies indicate that PRKCDBP binds not only to PKC-delta but also to caveolin-1 and helps regulate caveolin traffic and function. Similar to other members of the cavin family, the expression of PRKCDBP was found to be down-regulated in various cancer cell lines, suggesting a possible tumor suppressor function of PRKCDBP.Specifications
SRBC | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PRKCDBP; protein kinase C, delta binding protein; HSRBC; MGC20400; SRBC; sdr-related gene product that binds to c-kinase; cavin-3 | |
CAVIN3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q969G5 | |
112464 | |
Between 225 and 250 | |
Primary | |
4° C | |
CAVIN3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.