Learn More
Abnova™ SRC (Human) Recombinant Protein
For use with Functional Study.
Manufacturer:  Abnova™ P6561
Catalog No. 89-339-476
Add to cart
Description
Description
Human SRC (NP_005408.1, 1 a.a. - 536 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
Functional Study
v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
5 µg
ASV, SRC1, c-SRC, p60-Src
6714
The purity was assessed by SDS-PAGE/CBB staining.
Stored at -80°C. Aliquot to avoid repeated freezing and thawing.