Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Src Mouse anti-Avian, Human, Mouse, Rat, Mink, Clone: GD11, MilliporeSigma™

Mouse Monoclonal Antibody

$373.00

Specifications

Antigen Src
Clone GD11
Concentration 0.5 mg/mL
Applications Immunoprecipitation, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05184I100UL
View Documents
MilliporeSigma
05184I100UL
100 μL
Each for $373.00
Add to cart
 
05184I25UL
View Documents
MilliporeSigma
05184I25UL
25 μL
Each for $373.00
Add to cart
 
Description

Description

Src Monoclonal antibody specifically detects Src in Avian, Human, Mouse, Rat, Mink samples. It is validated for Immunoprecipitation, IP-Kinase Assay, Western Blotting.
Specifications

Specifications

Src
0.5 mg/mL
Unconjugated
Mouse
Avian, Human, Mouse, Rat, Mink
A recombinant fragment expressed in E.coli corresponding to 88 amino acids from the N-terminal half of human Proto-oncogene tyrosine-protein kinase Src (c-Src).
Monoclonal
Protein G purified
GD11
Immunoprecipitation, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit