Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC Mouse anti-Human, Clone: 5A18, Abnova™

Mouse monoclonal antibody raised against human SRC.

Manufacturer:  Abnova Corporation MAB12569

Catalog No. 89-934-963


Add to cart

Description

Description

This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

Specifications

SRC
5A18
Mouse monoclonal antibody raised against human SRC.
In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide).
ASV/SRC1/c-SRC/p60-Src
Mouse
Protein G purification
RUO
Primary
6714
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
SRC
Recombinant protein corresponding to full length human SRC.
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit