Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRD5A2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen SRD5A2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159525
SDP
View Documents
Novus Biologicals
NBP159525
100 μL
Each of 1 for $482.50
Add to Cart
 
Description

Description

SRD5A2 Polyclonal specifically detects SRD5A2 in Human, Monkey samples. It is validated for Western Blot.
Specifications

Specifications

SRD5A2
Polyclonal
Rabbit
Cancer
3-oxo-5-alpha-steroid 4-dehydrogenase 2,5 alpha-SR2, EC 1.3.99.5, MGC138457, S5AR 2, SR type 2, Steroid 5-alpha-reductase 2,3-oxo-5 alpha-steroid 4-dehydrogenase 2, steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 2), Type II 5-alpha reductase
SRD5A2
Primary
Western Blot
Unconjugated
RUO
P31213
6716
IgG
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit