Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREB3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen SREB3
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124119
View Documents
Novus Biologicals
NBP309609100UL
100 μg
Each for $352.89
Add to cart
 
NB124120
View Documents
Novus Biologicals
NBP30960925UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

SREB3 Polyclonal antibody specifically detects SREB3 in Human samples. It is validated for Western Blot
Specifications

Specifications

SREB3
Western Blot
Purified
RUO
Human
G protein coupled receptor 173, G protein-coupled receptor 173, G-protein coupled receptor 173, probable G-protein coupled receptor 173, SREB3Super conserved receptor expressed in brain 3
The immunogen is a synthetic peptide directed towards the C-terminal region of Human SREB3 (NP_061842). Peptide sequence IRQNGHAASRRLLGMDEVKGEKQLGRMFYAITLLFLLLWSPYIVACYWRV
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
GPCR
PBS buffer, 2% sucrose
54328
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit