Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Mouse anti-Human, Clone: SREBP1/1578, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30787810UG

 View more versions of this product

Catalog No. NB123229


Add to cart

Description

Description

SREBP1 Monoclonal antibody specifically detects SREBP1 in Human samples. It is validated for ELISA
Specifications

Specifications

SREBP1
SREBP1/1578
Unconjugated
10 mM PBS with 0.05% BSA
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
Recombinant full-length human SREBP1 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
ELISA
0.2 mg/ml
ELISA
Purified
Mouse
IgG2b Kappa
100 μg
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primary
6720
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit