Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SREBP1 Mouse anti-Human, Clone: SREBP1/1578, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | SREBP1 |
---|---|
Clone | SREBP1/1578 |
Concentration | 0.2 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123230
|
Novus Biologicals
NBP30787820UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
SREBP1 Monoclonal specifically detects SREBP1 in Human samples. It is validated for ELISA.Specifications
SREBP1 | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
6720 | |
IgG2b κ | |
Protein A or G purified |
SREBP1/1578 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
Recombinant full-length human SREBP1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title