Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRGAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562604
Description
Immunogen sequence: SPTDRHPDGY LARQRKRGEP PPPVRRPGRT SDGHCPLHPP HALSNSSVDL GSPSLASHPR GLLQNRGLNN DSPERRRRPG HGSLTNISRH DSLKKIDSPP IRRSTSSGQY TGFND Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
The protein encoded by this gene is a GTPase activator, working with the GTPase CDC42 to negatively regulate neuronal migration. The encoded protein interacts with the transmembrane receptor ROBO1 to inactivate CDC42.
Specifications
SRGAP1 | |
Polyclonal | |
Unconjugated | |
SRGAP1 | |
4930572H05Rik; Arhgap13; KIAA1304; NMTC2; RGD1566260; Rho GTPase activating protein 13; rho GTPase-activating protein 13; SLIT-ROBO Rho GTPase activating protein 1; SLIT-ROBO Rho GTPase-activating protein 1; Srgap1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57522 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6B7 | |
SRGAP1 | |
Recombinant protein corresponding to Human SRGAP1. Recombinant protein control fragment (Product #RP-101995). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction