Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRGAP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548908

Catalog No. PA548908

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTPase-activating protein for RhoA and Cdc42 small GTPases. Together with CDC42 seems to be involved in the pathway mediating the repulsive signaling of Robo and Slit proteins in neuronal migration. SLIT2, probably through interaction with ROBO1, increases the interaction of SRGAP1 with ROBO1 and inactivates CDC42.
Specifications

Specifications

SRGAP1
Polyclonal
PBS with 0.09% sodium azide
Q7Z6B7, Q91Z69
SLIT-ROBO Rho GTPase-activating protein 1, srGAP1, Rho GTPase-activating protein 13, SRGAP1, ARHGAP13, KIAA1304
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
117600, 57522
Western Blot
Unconjugated
SRGAP1
Liquid
SRGAP1
KLH conjugated synthetic peptide between 469-497 amino acids from the Central region of human SRGAP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit