Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRGAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550888

Catalog No. PA550888


Add to cart

Description

Description

The antibody detects endogenous levels of total SRGAP2 protein.

This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
Specifications

Specifications

SRGAP2
Polyclonal
Unconjugated
SRGAP2
Liquid
SRGAP2
Synthetic peptide corresponding to a region derived from internal residues of human SLIT-ROBO Rho GTPase activating protein 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O75044, Q91Z67, D4A208
ARHGAP34, FNBP2, KIAA0456, SRGAP2A
Rabbit
IgG
100 μL
-20°C
Primary
14270, 23380, 360840
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit