Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRM Mouse anti-Human, Clone: 6.1_1B8, Abnova™

Mouse monoclonal antibody raised against human SRM.

Manufacturer:  Abnova Corporation MAB12757

Catalog No. 89-934-964

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. Spermidine synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor. [provided by RefSeq]

Specifications

Specifications

SRM
6.1_1B8
Mouse monoclonal antibody raised against human SRM.
In PBS (0.02% sodium azide).
PAPT/SPDSY/SPS1/SRML1
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunocytochemistry (10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
SRM
Human HeLa cells
Protein A/G purification
RUO
Primary
6723
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit