Learn More
SRP54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179761AP
Catalog No. 50-173-1912
Add to cart
Description
Description
This antibody specifically recognizes the 54 kDa SRP54 protein.
Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
Specifications
Specifications
SRP54
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14576, P61011, Q6AYB5
SRP54
Rabbit
IgG
150 μL
-20°C
Primary
116650, 24067, 6729
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SRP54
Liquid
SRP54
SRP54 Fusion Protein Ag12166
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat