Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRPIN340, Tocris Bioscience™
Click to view available options
Quantity:
10mg
50mg
Description
Specifications
Quantity | 10mg |
Color | White |
Formula Weight | 349.35 |
Physical Form | Solid |
Percent Purity | >98% |
Chemical Name or Material | SRPIN340 |
Safety and Handling
Recommended Storage : Store at +4°C