Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRPIN340, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective serine arginine protein kinase (SRPK) 1 inhibitor (Ki = 0.89 μM). Inhibits SRPK2 at higher concentrations. Does not significantly inhibit other SRPKs such as CLK1 and CLK4, or other classes of SR kinases. Down-regulates expression of VEGF165 but does not affect VEGF165b expression.
Specifications
Color | White |
Quantity | 50 mg |
Formula Weight | 349.35 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | SRPIN340 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction