Learn More
Aviva Systems Biology Corp SRPRB Antibody - C-terminal region (ARP49644_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49644_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SRPRB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SRPRB. Alternate names: APMCF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60134