Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRPX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533059
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Monkey, Bat, Rabbit (100%) Galago, Marmoset, Elephant, Panda, Dog, Horse, Pig (92%).
Sushi-repeat-containing protein X-linked 2 (SRPX2) is a neural gene functioning in the speech and language center of the human brain; mutations in this gene lead to epilepsy, speech dyspraxia, mental retardation and cognitive disorders. Recently, SRPX2 was found to be a novel mediator of angiogenesis and can act as a ligand for the urokinase-type plasminogen activator, a protein that can facilitate invasive migration of sprouting endothelial cells. SRPX2 is also overexpressed in gastric cancer, leading to increased phosphorylation levels of focal adhesion kinase and enhanced cellular migration and adhesion, suggesting that SRPX2 may be a potential target in the treatment of metastatic cancers.
Specifications
SRPX2 | |
Polyclonal | |
Unconjugated | |
SRPX2 | |
1110039C07Rik; BPP; CBPS; EMBL:AAI68975.1, ECO:0000312; PMGX; PubMed:24179158}; RESDX; RGD:1562444}; RGD1562444; RP11-524D16__A.1; SRP; SRPUL; SRPX2; srpx2 {ECO:0000312; sushi repeat containing protein X-linked 2; sushi repeat containing protein, X-linked 2; sushi repeat-containing protein SRPX2; sushi repeat-containing protein SRPX2 {ECO:0000303; sushi-repeat containing protein; sushi-repeat containing protein, X-linked 2; sushi-repeat protein; sushi-repeat protein upregulated in leukemia; sushi-repeat protein up-regulated in leukemia; sushi-repeat-containing protein, X-linked 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100343092, 27286 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 0.1% sodium azide | |
O60687 | |
SRPX2 | |
Synthetic 13 amino acid peptide from C-Terminus of human SRPX2. | |
50 μg | |
Primary | |
Human, Rabbit, Bat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction