Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine racemase Mouse anti-Human, Clone: OTI7E8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525058

Catalog No. PIMA525058


Add to cart

Description

Description

SRR catalyzes the synthesis of D-serine from L-serine.
Specifications

Specifications

Serine racemase
Monoclonal
0.66 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9GZT4
ILV1; ISO1; serine racemase; D-serine ammonia-lyase; D-serine dehydratase; L-serine ammonia-lyase; L-serine dehydratase
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
63826
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7E8
Unconjugated
SRR
Liquid
SRR
Full length human recombit protein of SRR produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit