Learn More
Aviva Systems Biology Corp SRRD Antibody - middle region (ARP48022_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRRD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SRRD. Alternate names: HC/HCC, srr1l, SRR1LApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-59704