Learn More
Analytical Sales & Services SS 0.2µm Ultra High Pressure Direct Connect Pre-Column Filter Guard; 15,000 Psi  DFS Item
Manufacturer:  Analytical Sales & Services 84555
SS 0.2um Ultra High Pressure Direct Connect Pre-Column Filter Guard; 15,000 PSI
Catalog No. 50-646-269