Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ SS-174 PTFE-Faced Septa
Suitable to overcome the degradation of the septa, due to prolonged high temperatures which occurs in silicone rubber septa
Supplier: MilliporeSigma™ Supelco™ 22647
Description
- Septa degradation results in baseline drift and additional peaks
- When the PTFE is faced towards the column packing this issue is eliminated
- Designed for applications where greater inertness is required
- The off-white silicone rubber body (123 mil) provides plenty of support for the yellow PTFE-face (2 mil)
- After the PTFE is ruptured during injection, some bleeding may occur
- It is recommend to use a needle guide to minimize the size of the punctured area
- For use with 200°C to 300 °C inlet temperatures

Specifications
10 mm | |
13/32 in. | |
300°C | |
Silicone |
123 mil | |
PTFE Face | |
SS-174 Septa | |
100 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction