Learn More
Thermo Scientific™ SS-34 Fixed Angle Rotor
Support a wide range of applications with the Thermo Scientific™ SS-34 Fixed Angle Rotor, which features eight places.
Supplier: Thermo Scientific™ 28020
Description
Support a wide range of applications with the Thermo Scientific™ SS-34 Fixed Angle Rotor, which features eight places.
Compatible with:
Thermo Scientific™ Sorvall™ RC-6 Plus Superspeed Centrifuge.
Recommended for:
Multi-liter cell culture harvesting or the collection of cell-free growth media containing excreted products such as proteins, hormones and vitamins.
Specifications
SS-34 Fixed Angle Rotor | |
Fixed Angle Rotor | |
50,228 x g | |
107 mm | |
714 | |
29 mm | |
Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges | |
15,350 x g |
Aluminum | |
8 x 50 mL | |
25,000 rpm | |
32.7 mm | |
108 mm | |
31.75 lb. (15 kg) | |
34° |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.