Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SS-A 60 IgG ELISA Kit

Enzyme-linked immunosorbent assay for the detection of IgG antibodies to SS-A 60

Manufacturer:  Abnova™ KA4868

Catalog No. 89-032-316


Add to cart

Description

Description

Intended for detection of specific IgG antibodies in a sample by means of a sandwich type of the EIA method (i.e. a solid phase coated with specific antigen – antibody from the analyzed sample – labeled antibody).
Specifications

Specifications

5 to 320 U/mL
SS-A 60 IgG ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgG antibodies to SS-A 60.
CUT-OFF (Calibrator 2): 20 U/mL (3mL)
For research use only (RUO)
Store the kit at 4°C. Do not freeze.
Colorimetric
Human
HRP
Qualitative; Quantification
SS-A 60 IgG ELISA Kit
100 μL (Prediluted)
Human
1 Kit
Plasma, Serum
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit