Learn More
International CrystalSupplier Diversity Partner SS VIAL W/SS PESTLE 0013-2489 DFS Item
Manufacturer:  International CrystalSupplier Diversity Partner 00132489
Stainless Steel Vial (1 in. h. x 1/2 in. dia) with stainless steel ball pestle (1/4 in. dia.), Wig-L-Bug accessory
Catalog No. NC0037588