Learn More
SSBP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155951AP
Catalog No. 50-173-0336
Add to cart
Description
Description
SSBP4 contains 1 LisH domain. The exact function of SSBP4 remains unknown.
Specifications
Specifications
SSBP4
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BWG4
SSBP4
Rabbit
IgG
150 μL
-20°C
Primary
170463, 364534, 76900
Western Blot
Unconjugated
SSBP4
Liquid
Ssbp4
SSBP4 Fusion Protein Ag7952
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat