Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSC5D Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548777

Catalog No. PA548777

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Binds to extracellular matrix proteins. Binds to pathogen-associated molecular patterns (PAMPs) present on the cell walls of Gram-positive and Gram-negative bacteria and fungi, behaving as a pattern recognition receptor (PRR). Induces bacterial and fungal aggregation and subsequent inhibition of PAMP-induced cytokine release. Does not possess intrinsic bactericidal activity. May play a role in the innate defense and homeostasis of certain epithelial surfaces.
Specifications

Specifications

SSC5D
Polyclonal
PBS with 0.09% sodium azide
A1L4H1
Soluble scavenger receptor cysteine-rich domain-containing protein SSC5D, Soluble scavenger protein with 5 SRCR domains, SSc5D, SSC5D
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284297
Western Blot
Unconjugated
SSC5D
Liquid
SSC5D
KLH conjugated synthetic peptide between 869-897 amino acids from the Central region of human SSC5D.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit