Learn More
SSEA-4 Mouse anti-Human, Mouse, Canine, Chicken, Primate, Rabbit, Alexa Fluor™ 750, Clone: MC-813-70, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP226644AF750
Catalog No. NP226644A70
Add to cart
Description
Description
SSEA-4 Monoclonal antibody specifically detects SSEA-4 in Human,Mouse,Canine,Chicken,Primate,Rabbit samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
SSEA-4
MC-813-70
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
Mouse
IgG3 Kappa
Store at 4°C in the dark.
Monoclonal
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Human embryonal carcinoma cell line 2102Ep
0.1 mL
Primary
Canine, Chicken, Human, Mouse, Primate, Rabbit