Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-5 Mouse anti-Human, Alexa Fluor™ 594, Clone: 800000000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242922AF594

Catalog No. NP242922A94


Add to cart

Description

Description

SSEA-5 Monoclonal antibody specifically detects SSEA-5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

SSEA-5
800000000000
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
Undifferentiated human embryonic stem cells
Protein G purified
Store at 4°C in the dark.
Monoclonal
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG
0.1 mL
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit