Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

SSEA-5 Mouse anti-Human, HRP, Clone: 800000000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242922H

Catalog No. NBP242922H



Description

Description

SSEA-5 Monoclonal antibody specifically detects SSEA-5 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

SSEA-5
800000000000
Immunohistochemistry
Affinity Purified
Undifferentiated human embryonic stem cells
Immunogen affinity purified
Store at 4C in the dark.
Monoclonal
Immunohistochemistry
HRP
PBS with No Preservative
Mouse
IgG
0.1 ml
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit