Learn More
SSEA4 Mouse anti-Human, Alexa Fluor® 700, Clone: MC813-70, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen SSEA429
Catalog No. 50-113-7618
Add to cart
Description
Description
SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
Specifications
Specifications
SSEA4
Monoclonal
Alexa Fluor® 700
Liquid
Mouse
IgG3
500 μL
4° C
Primary
Human
Flow Cytometry
MC813-70
PBS with 0.1% sodium azide
stage-specific embryonic antigen 4; SSEA-4
Human SSEA4 antigen
Affinity chromatography
RUO
Antibody
Monoclonal