Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSH1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549200

Catalog No. PA549200

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Protein phosphatase which regulates actin filament dynamics. Dephosphorylates and activates the actin binding/depolymerizing factor cofilin, which subsequently binds to actin filaments and stimulates their disassembly. Inhibitory phosphorylation of cofilin is mediated by LIMK1, which may also be dephosphorylated and inactivated by this protein.
Specifications

Specifications

SSH1
Polyclonal
PBS with 0.09% sodium azide
Q8WYL5
Protein phosphatase Slingshot homolog 1, SSH-like protein 1, SSH-1L, hSSH-1L, SSH1, KIAA1298, SSH1L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54434
Western Blot
Unconjugated
SSH1
Liquid
SSH1
KLH conjugated synthetic peptide between 227-254 amino acids from the N-terminal region of human SSH1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit